5-Chloro-6-(difluoromethyl)pyrimidin-4-ol

Lieferant: Apollo Scientific
Achtung

PC50327-100MG
APOSPC50327-100MGEA 21.9 EUR
APOSPC50327-100MG
5-Chloro-6-(difluoromethyl)pyrimidin-4-ol
5-Chloro-6-(difluoromethyl)pyrimidin-4-ol
Formel: C₅H₃ClF₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09909765
CAS-Nummer: 425394-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR