3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Tert-Butylcarbamoyl)-4-fluorophenylboronic acid

PC5034-1G
APOSPC5034-1GEA 306 EUR
APOSPC5034-1G
3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid 98%
3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,05 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235045
CAS-Nummer: 874219-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR