1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene 97%

Lieferant: Apollo Scientific
Achtung

PC50377-1G
APOSPC50377-1GEA 162 EUR
APOSPC50377-1G
1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene 97%
1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene
Formel: C₇H₅ClFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11847613
CAS-Nummer: 1089280-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR