4-(Difluoromethyl)-2-(trifluoromethyl)anisole 95%

Lieferant: Apollo Scientific
Gefahr

PC50391-1G PC50391-250MG
APOSPC50391-1GEA 89.9 EUR
APOSPC50391-1G APOSPC50391-250MG
4-(Difluoromethyl)-2-(trifluoromethyl)anisole 95%
4-(Difluoromethyl)-2-(trifluoromethyl)anisole
Formel: C₉H₇F₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD14698614
CAS-Nummer: 1214348-62-5
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR