3-Cyan-4-fluoranilin 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril

PC50393-100G PC50393-25G
APOSPC50393-100GEA 149 EUR
APOSPC50393-100G APOSPC50393-25G
3-Cyan-4-fluoranilin 95%
3-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
Lagertemperatur: Kühlschrank
MDL: MFCD00277872
CAS-Nummer: 53312-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR