Mangan(III)fluorid 99%
Lieferant: Apollo Scientific
|
Gefahr
|
PC5040-10G
APOSPC5040-10GEA
122
EUR
APOSPC5040-10G
Mangan(III)fluorid 99%
Mangan(III)fluorid
Formel:
F₃Mn Dichte: 3,54 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016223 CAS-Nummer: 7783-53-1 EINECS: 232-006-6 UN: 3087 ADR: 5.1+6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...