3-Carbamoyl-5-fluorobenzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Carbamoyl-5-fluorophenyl)boronic acid, 3-(Aminocarbonyl)-5-fluorophenylboronic acid
PC5042-1G
APOSPC5042-1GEA
285
EUR
APOSPC5042-1G
3-Carbamoyl-5-fluorobenzeneboronic acid 98%
3-Carbamoyl-5-fluorobenzeneboronic acid
Formel:
C₇H₇BFNO₃ Molecular Weight: 182,95 g/mol Schmelzpunkt: 280…286 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08056357 CAS-Nummer: 871332-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...