4-Fluoro-3-(trifluoromethyl)benzyl fluoride 98%

Lieferant: Apollo Scientific

PC50420-1G
APOSPC50420-1GEA 132 EUR
APOSPC50420-1G
4-Fluoro-3-(trifluoromethyl)benzyl fluoride 98%
4-Fluoro-3-(trifluoromethyl)benzyl fluoride
Formel: C₈H₅F₅
Molecular Weight: 196,117 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1609026-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR