4-Chloro-3-(trifluoromethoxy)bromobenzene 97%

Lieferant: Apollo Scientific

PC50436-10G PC50436-1G PC50436-5G PC50436-25G
APOSPC50436-10GEA 575 EUR
APOSPC50436-10G APOSPC50436-1G APOSPC50436-5G APOSPC50436-25G
4-Chloro-3-(trifluoromethoxy)bromobenzene 97%
4-Chloro-3-(trifluoromethoxy)bromobenzene
Formel: C₇H₃BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09878140
CAS-Nummer: 406232-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR