3-Chloro-5-(trifluoromethoxy)bromobenzene 97%

Lieferant: Apollo Scientific

PC50439-10G PC50439-1G PC50439-5G PC50439-25G
APOSPC50439-10GEA 575 EUR
APOSPC50439-10G APOSPC50439-1G APOSPC50439-5G APOSPC50439-25G
3-Chloro-5-(trifluoromethoxy)bromobenzene 97%
3-Chloro-5-(trifluoromethoxy)bromobenzene
Formel: C₇H₃BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11856023
CAS-Nummer: 1417567-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR