3-Methoxy-5-(trifluormethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Methoxy-5-trifluoromethylphenylboronic acid
PC5052-5G
APOSPC5052-5GEA
134
EUR
APOSPC5052-5G
3-Methoxy-5-(trifluormethyl)phenylboronsäure 98%
3-Methoxy-5-(trifluormethyl)phenylboronsäure
Formel:
C₈H₈BF₃O₃ Molecular Weight: 219.95 g/mol Schmelzpunkt: 159…162 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08056362 CAS-Nummer: 871332-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...