1-(2-Chloroethyl)-3-[4-(trifluoromethyl)phenyl]urea

Lieferant: Apollo Scientific
Achtung

PC50540-1G
APOSPC50540-1GEA 0 EUR
APOSPC50540-1G
1-(2-Chloroethyl)-3-[4-(trifluoromethyl)phenyl]urea
1-(2-Chloroethyl)-3-[4-(trifluoromethyl)phenyl]urea
Formel: C₁₀H₁₀ClF₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13225859
CAS-Nummer: 13908-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR