1-[2-Bromo-4-(trifluoromethyl)phenyl]imidazolidin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
PC50541-250MG
APOSPC50541-250MGEA
24.8
EUR
APOSPC50541-250MG
1-[2-Bromo-4-(trifluoromethyl)phenyl]imidazolidin-2-one
1-[2-Bromo-4-(trifluoromethyl)phenyl]imidazolidin-2-one
Formel:
C₁₀H₈BrF₃N₂O Molecular Weight: 309,083 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32264077 CAS-Nummer: 2407339-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...