1-[4-(Trifluoromethyl)phenyl]imidazolidin-2-one

Lieferant: Apollo Scientific
Achtung

PC50542-1G PC50542-250MG
APOSPC50542-1GEA 0 EUR
APOSPC50542-1G APOSPC50542-250MG
1-[4-(Trifluoromethyl)phenyl]imidazolidin-2-one
1-[4-(Trifluoromethyl)phenyl]imidazolidin-2-one
Formel: C₁₀H₉F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD22049301
CAS-Nummer: 162748-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR