1-[3-(Trifluoromethyl)phenyl]imidazolidin-2-one

Lieferant: Apollo Scientific
Achtung

PC50543-1G PC50543-250MG
APOSPC50543-1GEA 464 EUR
APOSPC50543-1G APOSPC50543-250MG
1-[3-(Trifluoromethyl)phenyl]imidazolidin-2-one
1-[3-(Trifluoromethyl)phenyl]imidazolidin-2-one
Formel: C₁₀H₉F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03787197
CAS-Nummer: 14088-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR