1,2-Diethynyl-3-fluorobenzene

Lieferant: Apollo Scientific

PC50550-5G
APOSPC50550-5GEA 2990 EUR
APOSPC50550-5G
1,2-Diethynyl-3-fluorobenzene
1,2-Diethynyl-3-fluorobenzene
Formel: C₁₀H₅F
Lagertemperatur: Kühlschrank
CAS-Nummer: 412041-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR