1,2-Diethynyl-4-fluorobenzene

Lieferant: Apollo Scientific

PC50551-5G
APOSPC50551-5GEA 3210 EUR
APOSPC50551-5G
1,2-Diethynyl-4-fluorobenzene
1,2-Diethynyl-4-fluorobenzene
Formel: C₁₀H₅F
Lagertemperatur: Kühlschrank
CAS-Nummer: 336106-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR