1,3-Diethynyl-4,6-difluorobenzene
Lieferant: Apollo Scientific
PC50554-5G
APOSPC50554-5GEA
2800
EUR
APOSPC50554-5G
1,3-Diethynyl-4,6-difluorobenzene
1,3-Diethynyl-4,6-difluorobenzene
Formel:
C₁₀H₄F₂ Molecular Weight: 162,136 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
1379822-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...