1,3-Diethynyl-5-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

PC50555-5G
APOSPC50555-5GEA 1690 EUR
APOSPC50555-5G
1,3-Diethynyl-5-(trifluoromethyl)benzene
1,3-Diethynyl-5-(trifluoromethyl)benzene
Formel: C₁₁H₅F₃
Lagertemperatur: Kühlschrank
CAS-Nummer: 1622871-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR