1,3-Diethynyl-5-(trifluoromethoxy)benzene

Lieferant: Apollo Scientific

PC50556-5G
APOSPC50556-5GEA 0 EUR
APOSPC50556-5G
1,3-Diethynyl-5-(trifluoromethoxy)benzene
1,3-Diethynyl-5-(trifluoromethoxy)benzene
Formel: C₁₁H₅F₃O
Lagertemperatur: Kühlschrank
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR