3-Ethynyl-2-fluorobenzonitrile

Lieferant: Apollo Scientific

PC50560-5G
APOSPC50560-5GEA 3930 EUR
APOSPC50560-5G
3-Ethynyl-2-fluorobenzonitrile
3-Ethynyl-2-fluorobenzonitrile
Formel: C₉H₄FN
Molecular Weight: 145,133 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 2043403-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR