3-Ethynyl-5-fluorobenzonitrile
Lieferant: Apollo Scientific
PC50562-5G
APOSPC50562-5GEA
2480
EUR
APOSPC50562-5G
3-Ethynyl-5-fluorobenzonitrile
3-Ethynyl-5-fluorobenzonitrile
Formel:
C₉H₄FN Lagertemperatur: Gefrierschrank |
MDL:
MFCD09832165 CAS-Nummer: 872122-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...