3-Chlor-5-nitrobenzotrifluorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC5062-1G
PC5062-25G
PC5062-5G
APOSPC5062-1GEA
21.9
EUR
APOSPC5062-1G
APOSPC5062-25G
APOSPC5062-5G
3-Chlor-5-nitrobenzotrifluorid 95%
3-Chlor-5-nitrobenzotrifluorid
Formel:
C₇H₃ClF₃NO₂ Siedepunkt: 206…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412202 CAS-Nummer: 401-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...