2-(Methylamino)benzotrifluorid 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-2-(trifluoromethyl)aniline, 2-(Trifluoromethyl)-N-methylaniline

PC5064G-1G
APOSPC5064G-1GEA 72.7 EUR
APOSPC5064G-1G
2-(Methylamino)benzotrifluorid 97%
2-(Methylamino)benzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,15 g/mol
Siedepunkt: 67…69 °C/20mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052196
CAS-Nummer: 14925-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR