1-(4-Aminophenyl)-3,3,3-trifluoropropan-1-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC50739-100MG
APOSPC50739-100MGEA
95.7
EUR
APOSPC50739-100MG
1-(4-Aminophenyl)-3,3,3-trifluoropropan-1-one 95%
1-(4-Aminophenyl)-3,3,3-trifluoropropan-1-one
Formel:
C₉H₈F₃NO Molecular Weight: 203,161 g/mol |
MDL:
MFCD19673474 CAS-Nummer: 7119-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...