3-(2,2,2-Trifluroacetamido)benzeneboronic acid 98%

Lieferant: Apollo Scientific

PC5078-1G
APOSPC5078-1GEA 176 EUR
APOSPC5078-1G
3-(2,2,2-Trifluroacetamido)benzeneboronic acid 98%
3-(2,2,2-Trifluroacetamido)benzeneboronic acid
Formel: C₈H₇BF₃NO₃
Schmelzpunkt: 290…293 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235083
CAS-Nummer: 88978-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR