4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-3-(hydroxymethyl)phenylboronic acid, 4-Fluoro-3-(hydroxymethyl)benzeneboronic acid

PC5081-1G PC5081-250MG PC5081-5G PC5081-10G PC5081-25G
APOSPC5081-1GEA 108 EUR
APOSPC5081-1G APOSPC5081-250MG APOSPC5081-5G APOSPC5081-10G APOSPC5081-25G
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid
Formel: C₇H₈BFO₃
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08235075
CAS-Nummer: 481681-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR