3-(Carboxymethoxy)-5-fluorobenzeneboronic acid 96%

Lieferant: Apollo Scientific
Achtung

PC5090-1G
APOSPC5090-1GEA 379 EUR
APOSPC5090-1G
3-(Carboxymethoxy)-5-fluorobenzeneboronic acid 96%
3-(Carboxymethoxy)-5-fluorobenzeneboronic acid
Formel: C₈H₈BFO₅
Molecular Weight: 213,956 g/mol
Schmelzpunkt: 230…232 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436015
CAS-Nummer: 913835-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR