4-(Ethylcarbamoyl)-3-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethylcarbamoyl-3-fluorobenzeneboronic acid

PC5091-1G
APOSPC5091-1GEA 176 EUR
APOSPC5091-1G
4-(Ethylcarbamoyl)-3-fluorphenylboronsäure 98%
4-(Ethylcarbamoyl)-3-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211,00 g/mol
Schmelzpunkt: 157…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436070
CAS-Nummer: 874289-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR