2-Chloro-4-(difluoromethyl)-1-iodobenzene 95%

Lieferant: Apollo Scientific

PC50917-1G PC50917-250MG
APOSPC50917-1GEA 0 EUR
APOSPC50917-1G APOSPC50917-250MG
2-Chloro-4-(difluoromethyl)-1-iodobenzene 95%
2-Chloro-4-(difluoromethyl)-1-iodobenzene
Formel: C₇H₄ClF₂I
Lagertemperatur: Raumtemperatur
MDL: MFCD18392844
CAS-Nummer: 1261821-31-1
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR