4-Fluor-3-(methoxy(methyl)carbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

PC5093-1G
APOSPC5093-1GEA 176 EUR
APOSPC5093-1G
4-Fluor-3-(methoxy(methyl)carbamoyl)phenylboronsäure 98%
4-Fluor-3-(methoxy(methyl)carbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
Schmelzpunkt: 160…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235106
CAS-Nummer: 874290-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR