5-(Butylcarbamoyl)-2-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-n-Butylcarbamoyl-2-fluorobenzeneboronic acid

PC5094-1G
APOSPC5094-1GEA 176 EUR
APOSPC5094-1G
5-(Butylcarbamoyl)-2-fluorphenylboronsäure 98%
5-(Butylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,05 g/mol
Schmelzpunkt: 174…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235104
CAS-Nummer: 874289-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR