1-Bromo-5-chloro-3,4-difluoro-2-iodobenzene 95%

Lieferant: Apollo Scientific

PC50949-1G PC50949-250MG PC50949-5G
APOSPC50949-1GEA 522 EUR
APOSPC50949-1G APOSPC50949-250MG APOSPC50949-5G
1-Bromo-5-chloro-3,4-difluoro-2-iodobenzene 95%
1-Bromo-5-chloro-3,4-difluoro-2-iodobenzene
Formel: C₆HBrClF₂I
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2451256-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR