2-Deoxy-2,2-difluoro-D-glucose
Lieferant: Apollo Scientific
PC50973-1G
APOSPC50973-1GEA
1090
EUR
APOSPC50973-1G
2-Deoxy-2,2-difluoro-D-glucose
2-Deoxy-2,2-difluoro-D-glucose
Formel:
C₆H₁₀F₂O₅ Molecular Weight: 200,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30062869 CAS-Nummer: 34245-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...