2-Fluoro-4-piperazinobenzoic acid

Lieferant: Apollo Scientific

PC510008-5G
APOSPC510008-5GEA 804 EUR
APOSPC510008-5G
2-Fluoro-4-piperazinobenzoic acid
2-Fluoro-4-piperazinobenzoic acid
Formel: C₁₁H₁₃FN₂O₂
Molecular Weight: 224,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11872787
CAS-Nummer: 1121613-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR