Methyl 5-Fluoro-2-(4-methylpiperazino)benzoate

Lieferant: Apollo Scientific

PC510021-5G
APOSPC510021-5GEA 713 EUR
APOSPC510021-5G
Methyl 5-Fluoro-2-(4-methylpiperazino)benzoate
Methyl 5-Fluoro-2-(4-methylpiperazino)benzoate
Formel: C₁₃H₁₇FN₂O₂
Molecular Weight: 252,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071007
CAS-Nummer: 1256633-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR