Ethyl 4,5-Difluoro-2-nitrobenzoate
Lieferant: Apollo Scientific
PC510023-25G
APOSPC510023-25GEA
252
EUR
APOSPC510023-25G
Ethyl 4,5-Difluoro-2-nitrobenzoate
Ethyl 4,5-Difluoro-2-nitrobenzoate
Formel:
C₉H₇F₂NO₄ Molecular Weight: 231.153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11042826 CAS-Nummer: 1214387-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...