Ethyl (1R,2S)-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Lieferant: Apollo Scientific
PC510025-1G
APOSPC510025-1GEA
397
EUR
APOSPC510025-1G
Ethyl (1R,2S)-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Ethyl (1R,2S)-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Formel:
C₁₅H₂₀FNO₂ Molecular Weight: 265.323 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651691 CAS-Nummer: 1033756-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...