Ethyl (1S,2R)-2-(4-Fluorobenzylamino)cyclopentanecarboxylate

Lieferant: Apollo Scientific

PC510026-1G
APOSPC510026-1GEA 357 EUR
APOSPC510026-1G
Ethyl (1S,2R)-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Ethyl (1S,2R)-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Formel: C₁₅H₂₀FNO₂
Molecular Weight: 265.323 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651693
CAS-Nummer: 1140972-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR