5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
Lieferant: Apollo Scientific
PC510029-5G
APOSPC510029-5GEA
1220
EUR
APOSPC510029-5G
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
Formel:
C₁₁H₁₁FN₂O Molecular Weight: 206,216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651727 CAS-Nummer: 1272756-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...