5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one

Lieferant: Apollo Scientific

PC510030-5G
APOSPC510030-5GEA 1120 EUR
APOSPC510030-5G
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
5-Fluorospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
Formel: C₁₂H₁₃FN₂O
Molecular Weight: 220.243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651732
CAS-Nummer: 1272756-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR