3-(4-Bromo-2-fluorophenyl)-1,4-diazaspiro[4.4]nonan-2-one

Lieferant: Apollo Scientific

PC510032-5G
APOSPC510032-5GEA 1120 EUR
APOSPC510032-5G
3-(4-Bromo-2-fluorophenyl)-1,4-diazaspiro[4.4]nonan-2-one
3-(4-Bromo-2-fluorophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Formel: C₁₃H₁₄BrFN₂O
Molecular Weight: 313,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651736
CAS-Nummer: 1272755-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR