7-(5-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one

Lieferant: Apollo Scientific

PC510034-5G
APOSPC510034-5GEA 1220 EUR
APOSPC510034-5G
7-(5-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
7-(5-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
Formel: C₁₂H₁₂BrFN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18651744
CAS-Nummer: 1272755-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR