2-Ethoxy-5-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific

PC510039-5G
APOSPC510039-5GEA 1330 EUR
APOSPC510039-5G
2-Ethoxy-5-(trifluoromethyl)benzaldehyde
2-Ethoxy-5-(trifluoromethyl)benzaldehyde
Formel: C₁₀H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06247416
CAS-Nummer: 472809-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR