2-Propoxy-5-(trifluoromethyl)benzaldehyde
Lieferant: Apollo Scientific
PC510040-5G
APOSPC510040-5GEA
1330
EUR
APOSPC510040-5G
2-Propoxy-5-(trifluoromethyl)benzaldehyde
2-Propoxy-5-(trifluoromethyl)benzaldehyde
Formel:
C₁₁H₁₁F₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06248747 CAS-Nummer: 472809-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...