6-[4-(Trifluoromethoxy)phenyl]nicotinic acid

Lieferant: Apollo Scientific

PC510041-5G
APOSPC510041-5GEA 1620 EUR
APOSPC510041-5G
6-[4-(Trifluoromethoxy)phenyl]nicotinic acid
6-[4-(Trifluoromethoxy)phenyl]nicotinic acid
Formel: C₁₃H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06410408
CAS-Nummer: 851266-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR