2-Amino-2-(4-bromo-2-fluorophenyl)acetic acid hydrochloride
Lieferant: Apollo Scientific
PC510042-5G
APOSPC510042-5GEA
1230
EUR
APOSPC510042-5G
2-Amino-2-(4-bromo-2-fluorophenyl)acetic acid hydrochloride
2-Amino-2-(4-bromo-2-fluorophenyl)acetic acid hydrochloride
Formel:
C₈H₇BrFNO₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371461 CAS-Nummer: 1136881-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...