2-(Hydroxymethyl)-6-[4-(trifluoromethyl)phenyl]pyridine

Lieferant: Apollo Scientific

PC510043-5G
APOSPC510043-5GEA 1540 EUR
APOSPC510043-5G
2-(Hydroxymethyl)-6-[4-(trifluoromethyl)phenyl]pyridine
2-(Hydroxymethyl)-6-[4-(trifluoromethyl)phenyl]pyridine
Formel: C₁₃H₁₀F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06410114
CAS-Nummer: 648439-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR