6-(3-Fluorphenyl)-3-pyridincarbaldehyd

Lieferant: Apollo Scientific

Synonyme: 6-(3-Fluorophenyl)nicotinaldehyde

PC510044-5G
APOSPC510044-5GEA 667 EUR
APOSPC510044-5G
6-(3-Fluorphenyl)-3-pyridincarbaldehyd
6-(3-Fluorphenyl)-3-pyridincarbaldehyd
Formel: C₁₂H₈FNO
Molecular Weight: 201.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115444
CAS-Nummer: 898795-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR