2-(3-Fluorophenyl)-5-(hydroxymethyl)pyridine

Lieferant: Apollo Scientific

PC510045-5G
APOSPC510045-5GEA 1540 EUR
APOSPC510045-5G
2-(3-Fluorophenyl)-5-(hydroxymethyl)pyridine
2-(3-Fluorophenyl)-5-(hydroxymethyl)pyridine
Formel: C₁₂H₁₀FNO
Molecular Weight: 203.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410430
CAS-Nummer: 887974-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR